TLC Analysis of Adenine, Adenosine and Their Phosphate Analogs on HPTLC NH2 F254s
TLC Analysis of Adenine, Adenosine and Their Phosphate Analogs on HPTLC NH2 F254s
Conditions
plate: 1.50451 HPTLC pre-coated plate NH2 F 254s (1.13192.0001. 1.15647.0001)
eluent: [A] ethanol; [B] water; (30:70, A:B) containing 0.2 M NaCl
migration distance: 5 cm
chamber: normal chamber without chamber saturation
detector: in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm.
sample: 0.3 μL
Description
suitability application for TLC