TLC Analysis of Aflatoxins on HPTLC Silica Gel 60 F 254 with Concentrating Zone
TLC Analysis of Aflatoxins on HPTLC Silica Gel 60 F 254 with Concentrating Zone
Conditions
plate  HPTLC pre-coated plate silica gel 60 F254 with concentrating zone
(1.13728.0001, 1.13727.0001)
eluent [A] chloroform; [B] acetone; (90:10, A:B)
migration distance  7 cm
chamber  normal chamber without chamber saturation
sample  1 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 334 nm
Description
Featured Industry Food and Beverages
suitability application for TLC