TLC Analysis of Alkaloids on HPTLC Silica Gel 60 F 254
TLC Analysis of Alkaloids on HPTLC Silica Gel 60 F 254
Conditions
plate: 1.05642 HPTLC pre-coated plate silica gel 60 F 254s (1.05642.0001, 1.05548.0001)
eluent: [A] methanol; [B] ammonia 25%; (99:1, A:B) migration distance: 7 cm
chamber: normal chamber without chamber saturation
detector: in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm suitability application for TLC Featured Industry Clinical