TLC Analysis of Amino Acids on HPTLC Pre-coated Cellulose without Fluorescent Indicator
TLC Analysis of Amino Acids on HPTLC Pre-coated Cellulose without Fluorescent Indicator
Conditions
plate HPTLC pre-coated plate cellulose without fluorescent indicator (1.16092.0001)
eluent [A] l-propanol; [B] water; (80:20, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
detector spray reagent: ninhydrin heated up to 110° C for 5 min
Description
Featured Industry Life Science and Biopharma
suitability application for TLC