TLC Analysis of Anabolic Compounds on HPTLC DIOL F254s (analyte set 2)
TLC Analysis of Anabolic Compounds on HPTLC DIOL F254s (analyte set 2)
Conditions
plate HPTLC pre-coated plaate DIOL F 254s (1.05636.0001; 1.12668.0001)
eluent [A] di-iso-propyl ether; [B] acetic acid glacial; (100:1, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
detector Spray reagent: MnCl2 – sulfuric acid heated up to 110° C for 5 min
in situ evaluation with TLC/HPTLC scanner (Camag) UV 366 nm
Description
suitability application for TLC