Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Analgesics on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
TLC Analysis of Analgesics on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
Conditions
sample 0.2 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 248 nm
plate HPTLC pre-coated plate RP-18 WF 254s (1.13124.0001)
migration distance 5 cm
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B] water; (40:60, A:B)
Description
Categories Analytical Chromatography, TLC Applications, analgesics
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13124.0001
suitability application for TLC