TLC Analysis of Antibiotics on HPTLC pre-coated plate silica 60 F 254s
TLC Analysis of Antibiotics on HPTLC pre-coated plate silica 60 F 254s
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s extra pure (suitable for aqueous eluents)
eluent water with addition of 2% adipic acid
migration distance 10 cm
chamber normal chamber without chamber saturation
detection in-situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma
Application No. BOR200918