TLC Analysis of Asparagine and Aspartic acid on HPTLC Silica 60F 254s
TLC Analysis of Asparagine and Aspartic acid on HPTLC Silica 60F 254s
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s extra pure (suitable for aqueous eluents)
eluent ethanol/buffer solution pH 3 85/15 (v/v)
with addition of 0,1 mol/1 tetramethylammonium chloride
migration distance 5 cm
chamber normal chamber without chamber saturation
sample application volume 300 nL
detection spray reagent: ninhydrin
heating up to 110 C for 5 min in-situ evaluation with TLC/HPTLC scanner (Camag) 500nm
Description
Featured Industry Life Science and Biopharma 
Analysis Note 0
Application No. BOR190918A