Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Benzoic Acid and Sorbic Acid on HPTLC CN F254s (analyte set 2)
TLC Analysis of Benzoic Acid and Sorbic Acid on HPTLC CN F254s (analyte set 2)
Conditions
sample 0.2 µL
detector Spray reagent: MnCI2- sulfuric acid heated up to 110 °C for 5 min in situ evaluation
with TLC/HPTLC scanner (Camag) UV 366 nm
eluent [A] petroleum benzine 40-60°; [B] acetone; (65:35, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
suitability application for TLC