TLC Analysis of Biphenylols on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
TLC Analysis of Biphenylols on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
Conditions
plate: TLC pre-coated plate silica gel 60 F 254 with concentrating zone (1.11798.0001)
eluent: [A] toluene; [B] chloroform; [C] methanol; (80:10:10, A:B:C) migration distance: 10 cm
chamber: normal chamber without chamber saturation
sample: 5 μL
detector: in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm suitability application for TLC Featured Industry Chemicals and Industrial Polymers