TLC analysis of Catecholamines by ion-pair chromatography on TLC pre-coated plate RP-8 F 254s
TLC analysis of Catecholamines by ion-pair chromatography on TLC pre-coated plate RP-8 F 254s
Conditions
plate TLC pre-coated plate RP-8 F 254s (P/N: 1.15389, Alternative P/N: 1.15423)
eluent Acetone/water 40/60 (v/v) with additio~ of 0,1 mol/l butane-l sodium sulfonate
migration distance 7 cm
chamber normal chamber without chamber saturation
detection in situ-evaluation with TLC/HPTLG scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma 
Analysis Note 0
Application No. BOR170918C