TLC Analysis of Chlorinated Hydrocarbons on HPTLC DIOL F254s
TLC Analysis of Chlorinated Hydrocarbons on HPTLC DIOL F254s
Conditions
sample 0.5 μL
eluent [A] petroleum benzine (40-60°)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate DIOL F254s (1.12668.0001)
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 220 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC