TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-18 F 254s (analyte set 2)
TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-18 F 254s
Conditions
sample 0.3 μL
detector dipped in dichlorofluorescein (0.2% in ethanol) (in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm)
eluent [A] acetonitrile; [B] water; (70:30, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate TLC pre-coated plate RP-18 F 254s (1.15389.0001, 1.15423.0001)
Description
Featured Industry
Vitamins, Nutraceuticals, and Natural Products
suitability application for TLC