TLC Analysis of Chlorophenols on HPTLC Pre-Coated Plate RP-8 F254s
TLC Analysis of Chlorophenols on HPTLC Pre-Coated Plate RP-8 F254s
Conditions
sample 0.3 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
eluent [A] methanol; [B] water; (65:35, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
suitability application for TLC