TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
sample 2 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
eluent [A] methanol; [B] water; (90:10, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate RP-18 F 254s with concentrating zone (1.15498.0001)
Description
suitability application for TLC