TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-2 F254s (analyte set 2)
TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-2 F254s (analyte set 2)
Conditions
plate  1.05747 HPTLC pre-coated plate RP-2 F 254s (1.05747.0001)
eluent [A] methanol; [B] water; (70:30, A:B) containing 0.1 M tetraethylammonium bromide
migration distance  7 cm
chamber  normal chamber without chamber saturation
sample  0.3 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers