Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Cholesterol and its Bile Acid Metabolites on HPTLC Pre-Coated Plate RP-18 F254s
TLC Analysis of Cholesterol and its Bile Acid Metabolites on HPTLC Pre-Coated Plate RP-18 F254s
Conditions
sample 0.2 µL
eluent [A] acetonitrile; [B] buffer solution pH 2.00; (50:50, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Food and Beverages
General description TLC Plate Information: 1.16464.0001
suitability application for TLC