TLC Analysis of Cholesteryl Esters on HPTLC Si 60 F254 with Concentrating Zone
TLC Analysis of Cholesteryl Esters on HPTLC Si 60 F254 with Concentrating Zone
Conditions
plate  HPTLC pre-coated plate silica gel 60 F254 with concentrating zone
(1.13728.0001, 1.13727.0001)
eluent [A] toluene; [B] petroleum benzene 40-60 °C; (40:60, A:B)
migration distance  7 cm
chamber  normal chamber without chamber saturation
sample  5 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 366 nm. (spray reagent
Description
Featured Industry Life Science and Biopharma
suitability application for TLC