TLC Analysis of Cholesteryl Esters on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
TLC Analysis of Cholesteryl Esters on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 2)
Conditions
sample 5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) 366 nm (spray reagent: MnCl2-sulfuric acid heated up to 110° C for 5 min)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC