TLC Analysis of Compounds of Aniline on HPTLC CN F254s
TLC Analysis of Compounds of Aniline on HPTLC CN F254s
Conditions
sample 0.1 μL
eluent [A] n-hexane; [B] toluene; [C] acetone; (70:25:5, A:B:C)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 238 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC