TLC Analysis of Cytosine, Cytidine and its Phosphates on HPTLC NH2 F254s
TLC Analysis of Cytosine, Cytidine and its Phosphates on HPTLC NH2 F254s
Conditions
eluent [A] acetone; [B] water; (30:70, A:B) containing 0.2 M NaCl
chamber normal chamber without chamber saturation
migration distance 5 cm
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
Description
suitability application for TLC