Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Dansyl Amino Acids on HPTLC CN F254s (analyte set 1)
TLC Analysis of Dansyl Amino Acids on HPTLC CN F254s (analyte set 1)
Conditions
sample 0.4 μL
detector
dipped in dichlorofluorescein (0.2% in ethanol) (in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm)
eluent [A] ethanol; [B] acetone; [C] water; (25:30:45, A:B:C) containing 0,2 M lithium chloride
chamber normal chamber without chamber saturation
migration distance 8 cm
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC