TLC Analysis of Dansyl Amino Acids on HPTLC CN F254s (analyte set 2)
TLC Analysis of Dansyl Amino Acids on HPTLC CN F254s (analyte set 2)
Conditions
sample 0.2 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 366 nm
eluent [A] methanol; [B] l-propanol; [C] water; (30:20:50, A:B:C) containing 0,1 M lithium chloride
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
Description
suitability application for TLC