TLC Analysis of Derivatives of Urea on HPTLC DIOL F254s
TLC Analysis of Derivatives of Urea on HPTLC DIOL F254s
Conditions
sample 0.2 μL
detector
dipped in dichlorofluorescein (0.2% in ethanol) (in situ evaluation with TLC/HPTLC scanner (Camag) UV 238 nm)
eluent [A] petroleum benzine (40-60°); [B] acetone; (80:20, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate DIOL F254s (1.12668.0001)
Description
suitability application for TLC