TLC Analysis of Derivatives of Urea on HPTLC NH2 F254s
TLC Analysis of Derivatives of Urea on HPTLC NH2 F254s
Conditions
sample 0.150 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 238 nm
eluent [A] chloroform; [B] toluene; (80:20, A:B)
chamber normal chamber without chamber saturation
migration distance 7 cm
plate HPTLC pre-coated plate NH2F 254s (1.13192.0001, 1.15647.0001)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC