TLC Analysis of Digested Proteins on HPTLC Silica gel 60 RP-8 F254s
TLC Analysis of Digested Proteins on HPTLC Silica gel 60 RP-8 F254s
Conditions
plate HPTLC Silica gel 60 RP-8 F254s (Product No. 1.13725)
Mobile phase methanol/water (70/30) + 0,1mol/l ammonium acetate
migration distance 5 cm
chamber normal chamber without chamber saturation
detection 0,02% fluorescamin solution in acetone spray 20s and dry 10min at room temperature UV @ 366nm
Description
Application No. BOR050918A