TLC Analysis of Digested Proteins on Silica Gel 60
TLC Analysis of Digested Proteins on Silica Gel 60
Conditions
plate ProteoChrom® HPTLC Silica gel 60 F254s (Product No. 1.05650)
Mobile phase
2-butanol/pyridin/ammonia25%/water (39/34/10/26)
migration distance 5 cm
chamber normal chamber without chamber saturation
detection 0,5% Ninhydrin spray 20s and heat 1-2 min at 110°C
Detection white light
Description
Application No. BOR200918D