TLC Analysis of Digested Proteins on Silica gel 60 RP-18
TLC Analysis of Digested Proteins on Silica gel 60 RP-18
Conditions
plate HPTLC Silica gel 60 RP-18 F254s (Product No. 1.13724)
Mobile phase
methanol/water (70/30) + 0,1mol/l ammonium acetate
migration distance 5 cm
chamber normal chamber without chamber saturation
detection 0,02% fluorescamin solution in acetone spray 20s and dry 10min at roomtemperature Detection UV @ 366nm
Description
Application No. BOR050918B