TLC Analysis of Dinitrophenols on HPTLC Pre-Coated Plate CN F254s (analyte set 1)
TLC Analysis of Dinitrophenols on HPTLC Pre-Coated Plate CN F254s (analyte set 1)
Conditions
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
eluent [A] acetone; [B] water; (40:60, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.1 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers