TLC Analysis of Gallic Acid and Gallic Acid Esters on HPTLC Silica Gel 60 RP18 WF254s
TLC Analysis of Gallic Acid and Gallic Acid Esters on HPTLC Silica Gel 60 RP18 WF254s
Conditions
plate HPTLC precoated plate RP18 w F 254s (1.13124.0001)
eluent [A] methanol; [B] 1N acetic acid; (30:70, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
  in situ evaluation with TLC/HPTLC scanner
(Camag) UV 265 nm
Description
Categories Analytical/Chromatography Applications, TLC Applications, organic acids
Featured Industry Vitamins, Nutraceuticals, and Natural Products
suitability application for TLC