TLC Analysis of Herbicides / Fungicides on HPTLC DIOL F254s
TLC Analysis of Herbicides / Fungicides on HPTLC DIOL F254s
Conditions
plate 1.05636 HPTLC pre-coated plate DIOL F 254s (1.05636.0001, 1.12668.0001 )
eluent [A] petroleum benzine (40 - 60°); [B] acetone; (85:15, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 238 nm
Description
suitability
application for TLC
Featured Industry Agriculture