TLC Analysis of Hormones on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
TLC Analysis of Hormones on HPTLC Silica Gel 60 RP-18 WF254s (analyte set 2)
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
eluent [A] acetonitrile; [B] water; (60:40, A:B)
migration distance 5 cm
sample 0.2 μL
detector Spray reagent: MnC12- slufuric acid heated up to 110°C for 5 min (in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC