TLC Analysis of Hydroxycinnamic Acids on HPTLC Silica Gel 60 F 254 with Concentrating Zone
TLC Analysis of Hydroxycinnamic Acids on HPTLC Silica Gel 60 F 254 with Concentrating Zone
Conditions
plate  HPTLC pre-coated plate silica gel 60 F254 with concentraling zone
(1.13728.0001, 1.13727.0001)
eluent [A] acetonitrile; [B] methanol; [C] water; (90:5:5, A:B:C)
migration distance  6 cm
chamber  normal chamber without chamber saturation
sample  1 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC