TLC Analysis of Ingredients in Analgesic Agents on HPTLC pre-coated Plate 60 RP-18 WF 254s with concentrating zone
TLC Analysis of Ingredients in Analgesic Agents on HPTLC pre-coated Plate 60 RP-18 WF 254s with concentrating zone
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
eluent [A] acetonitrile; [B] water; (40:60, A:B)
migration distance 6 cm
chamber normal chamber without chamber saturation
sample 5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) 248 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC