TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 2)
TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 2)
Conditions
sample 5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm
eluent [A] acetonitrile; [B] water; (40:60, A:B) containing 0.1 M ammonium bromide
Plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone(with 1.3124)
migration distance 7cm
chamber normal chamber without chamber saturation
Description
Featured Industry Life Science and Biopharma
suitability application for TLC