TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 4)
TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 4)
Conditions
plate 1.05628 HPTLC Silica gel 60 F 254 10 cm x 10 cm (1.05628.0001)
eluent [A] chloroform; [B] acetone; (9:1, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
detector UV 366 nm and scan with TLC Scanner III (CAMAG)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC