TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 3)
TLC Analysis of Ingredients in Analgesic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 3)
Conditions
plate TLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
eluent [A] acetonitrile; [B] water; (50:50, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 5 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC