TLC Analysis of Ingredients in Anti-Influenza Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 2)
TLC Analysis of Ingredients in Anti-Influenza Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 2)
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s with concentraling zone (product number 1.13124.0001)
eluent [A] methanol; [B] water; (50:50, A:B) containing 0.1 M ammonium bromide
migration distance 7 cm
sample 5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC