TLC Analysis of Ingredients in Antirheumatic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 1)
TLC Analysis of Ingredients in Antirheumatic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone (analyte set 1)
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
eluent [A] acetonitrile; [B] water; (40:60, A:B) containing 0.1 M ammonium bromide
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 5 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC