TLC Analysis of Ingredients in Broncholytic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone
TLC Analysis of Ingredients in Broncholytic Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
eluent [A] acetonitrile; [B] water; (30:70, A:B) containing 0.1 M TEACI
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 5 μL (10 mg/5 mL)
detector (in situ evaluation with TLC/HPTLC scanner (Camag) UV 265)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC