TLC Analysis of Ingredients in Dermatological Agents on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Ingredients in Dermatological Agents on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s ()
eluent [A] chloroform; [B] methanol; (95:5, A:B)
migration distance 5 cm
chamber normal chamber without chamber saturation
sample 0.2 μL 
spray reagent MnCl2 - sulfuric acid heated to 100 °C for 10 min,
in situ evaluation with TLC/HPTLC scanner (Camag), 500 nm
Description
suitability
application for TLC