TLC Analysis of Lipophilic Dye on HPTLC Silica gel 60 F254
TLC Analysis of Lipophilic Dye on HPTLC Silica gel 60 F254
Conditions
plate HPTLC Silica gel 60 F254, 20x10cm (1. 05642.0001)
article number 1. 05642.0001
mobile phase Toluene
migration distance 5 cm
chamber normal chamber without chamber saturation
sample application volume 2 µL
detection White light
Chromatographic Data
No. Compounds Concentration (mg/ml) Solvent
1 to 7 p-Methoxyazobenzol (52) 3.0 toluene
  Sudan II (79) 3.0 toluene
  Sudanviolett (89) 3.0 toluene
  Ceresviolett BRN (55) 1.0 toluene
  Hrf observed from these dyes 14, 26, 41, 59,71, and 82  
Description
Featured Industry Chemicals and Industrial Polymers
Application No. BOR210918A