Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Morphine Derivatives by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Morphine Derivatives by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nmdipped in
dichlorofluorescein (0.2% in ethanol)
plate TLC pre-coated plate RP-18 F 254s (1.15389.0001, 1.15423.0001)
migration distance 7 cm
chamber normal chamber without chamber saturation
eluent [A] acetone; [B] water; (40:60, A:B) containing 0.1 M heptanesulfonic acid sodium salt
Description
Categories Analytical Chromatography, TLC Applications, drugs of abuse
Featured Industry Pharmaceutical (small molecule)
General description TLC Plate Information: 1.15389.0001
suitability application for TLC