TLC Analysis of Mycotoxins on HPTLC Silica 60F 254s
TLC Analysis of Mycotoxins on HPTLC Silica 60F 254s
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s extra pure (suitable for aqueous eluents)
eluent chloroform/acetonitrile/ethanol/acetic acid glacial 80/10/10/1 (v/v)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample application volume 200 nL
detection in-situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Agriculture 
Application No. BOR190918D