TLC Analysis of N-Methyl Xanthine Derivatives on HPTLC NH2 F254s
TLC Analysis of N-Methyl Xanthine Derivatives on HPTLC NH2 F254s
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001 )
eluent [A] methanol; [B] acetonitrile; [C] chloroform; (10:80:10, A:B:C)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
suitability application for TLC