TLC Analysis of Nucleobases on HPTLC Silica Gel 60 RP-2 F254s (analyte set 2)
TLC Analysis of Nucleobases on HPTLC Silica Gel 60 RP-2 F254s (analyte set 2)
Conditions
plate  1.05747 HPTLC pre-coated plate RP-2 F 254s (1.05747.0001)
eluent [A] acetonitrile; [B] water; (60:40, A:B)
migration distance  7 cm
chamber  normal chamber without chamber saturation
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC