TLC Analysis of Nucleosides on HPTLC Silica 60 F254s
TLC Analysis of Nucleosides on HPTLC Silica 60 F254s
Conditions
plate HPTLC pre-coated plate silica gel 60 F 254s extra pure (suitable for aqueous eluents)
eluent
water
migration distance 7 cm
chamber normal chamber without chamber saturation
application volume 200 nL
detection in-situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Application No. BOR190918B