TLC Analysis of Nucleotides and Oligonucleotides on HPTLC NH2 F254s
TLC Analysis of Nucleotides and Oligonucleotides on HPTLC NH2 F254s
Conditions
plate HPTLC pre-coated plate NH2 F254s (1.15647.0001)
eluent [A] methanol; [B] water; (90:10, A:B) containing 0.2 M lithium chloride
migration distance 5 cm
chamber normal chamber without chamber saturation
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC