TLC Analysis of Oligonucleotides on HPTLC NH2 F254s (analyte set 2)
TLC Analysis of Oligonucleotides on HPTLC NH2 F254s (analyte set 2)
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001 )
eluent [A] ethanol; [B] water; (60:40, A:B) containing 0.2 M lithium chloride
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC