TLC Analysis of Pentapeptides on HPTLC Silica Gel 60 RP-18 F254s (analyte set 2)
TLC Analysis of Pentapeptides on HPTLC Silica Gel 60 RP-18 F254s (analyte set 2)
Conditions
sample 0.7 μL
detector (spray reagent: ninhydrin heated up to 1200 °C for 10 min)
eluent [A] acetonitrile; [B] 1N hydrochloric acid; (90:10, A:B) containing 0.1 M tetramethylammonium chloride
chamber normal chamber without chamber saturation
migration distance 8 cm
plate TLC pre-coated plate RP-18 F 254s (1.15389.0001, 1.15423.0001)
Description
Featured Industry Life Science and Biopharma
suitability application for TLC